missing translation for 'onlineSavingsMsg'
Learn More

Syntaxin Antibody (SP6) - Chimeric - Azide and BSA Free, Novus Biologicals™

Product Code. 18626568 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
Unit Size:
0.2mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18626568 0.2 mg 0.2mg
18619267 0.025 mg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18626568 Supplier Novus Biologicals Supplier No. NBP3089990.2mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Syntaxin Monoclonal antibody specifically detects Syntaxin in Human, Rat, Porcine, Bovine, Canine, Guinea Pig, Hamster, Rabbit samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Syntaxin
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone SP6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS
Gene Alias HPC-1;Sap;STX1A
Host Species Rabbit
Immunogen Synaptic vesicle-containing fractions immunoprecipitated from human brain homogenates.
Purification Method Protein A purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Neurodegeneration
Primary or Secondary Primary
Target Species Human, Rat, Pig, Bovine, Canine
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.