missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntaxin Antibody (SP6) - Chimeric - Azide and BSA Free, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Syntaxin Monoclonal antibody specifically detects Syntaxin in Human, Rat, Porcine, Bovine, Canine, Guinea Pig, Hamster, Rabbit samples. It is validated for Western Blot, Immunohistochemistry
Specifications
Specifications
| Antigen | Syntaxin |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | SP6 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS |
| Gene Alias | HPC-1;Sap;STX1A |
| Host Species | Rabbit |
| Immunogen | Synaptic vesicle-containing fractions immunoprecipitated from human brain homogenates. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?