Learn More
Syngene™ Long Wave UV Tube
Alle ansehen Syngene Produkte
Click to view available options
Wavelength:
365 nm
Packungsgröße:
Each
Beschreibung
- For use with all GX and GE model transilluminators
- 365nm wavelength
- Long wave
Spezifikation
Spezifikation
| Wavelength | 365 nm |
| For Use With (Equipment) | All GX/GE model transilluminators |
| Type | Longwave UV Tube |