missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Synapsin I Polyclonal specifically detects Synapsin I in Human, Mouse, Rat, Guinea Pig, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating.
Specifications
Specifications
| Antigen | Synapsin I |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:1000, Immunoprecipitation 1 ug per 200ug lysate, Immunohistochemistry-Frozen 1:10-1:500, Immunohistochemistry Free-Floating |
| Formulation | 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative |
| Gene Accession No. | P17599, P17600 |
| Gene Alias | SYN1a, SYN1b, Synapsin 1, Synapsin I, synapsin IBrain protein 4.1, synapsin-1, synaptic marker, synaptic markers immunohistochemistry, SYNI |
| Gene Symbols | SYN1 |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?