Learn More
Invitrogen™ Synapsin 1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ OPA104001
Description
In Western blot, this antibody detects an ~77 kDa and ~80 kDa protein representing synapsin I alpha and synapsin I beta respectively, in rat brain homogenate. Immunohistochemical staining of synapsin 1 alpha and 1 beta in human brain with OPA1-04001 yields a pattern consistent with cytoplasmic vesicle staining.
SYN1 (synapsin 1) is a member of the synapsin gene family. Synapsins encode neuronal phosphoproteins that associate with the cytoplasmic surface of synaptic vesicles. Family members are characterized by common protein domains and are implicated in synaptogenesis and the modulation of neurotransmitter release, suggesting a potential role in several neuropsychiatric diseases. Synapsin 1 plays a role in regulation of axonogenesis and synaptogenesis. It serves as a substrate for several different protein kinases and phosphorylation may function in the regulation of this protein in the nerve terminal. Mutations in this gene may be associated with X-linked disorders with primary neuronal degeneration such as Rett syndrome. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
| Synapsin 1 | |
| Polyclonal | |
| Unconjugated | |
| SYN1 | |
| Brain protein 4.1; SYN1; Syn-1; SYN1a; SYN1b; Syn1-S; synapsin 1; synapsin I; synapsin Ia; synapsin Ib; synapsin-1; SYNI; truncated synapsin 1-S | |
| Rabbit | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat, Bovine | |
| Antibody | |
| IgG |
| Western Blot | |
| Conc. Not Determined | |
| whole serum with no preservative | |
| O88935, P09951, P17599, P17600 | |
| SYN1 | |
| Native protein purified from bovine brain. | |
| RUO | |
| 20964, 24949, 281510, 6853 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.