Learn More
Thermo Scientific™ SWAP-IT™ GC-MS Interface System Accessories
Shop All Thermo Scientific ProductsDescription
Find the right nut and ferrule to use with the powerful Thermo Scientific™ SWAP-IT™ GC-MS Interface system.
- Specially deactivated , robust stainless steel transfer capillary provides an interface between the analytical column and the MS detector
- Small ID of transfer capillary ensures exposure to air and water is reduced to a minimum during disconnection of the analytical column
- The system works without the requirement for additional He-purge/make-up gas flow
- Finger-tight connections make exchanging the analytical column quick and easy without the need for tools
- Includes finger-tight connections for S/SL injector to provide complete finger-tight solution
Specifications
Specifications
| Type | Transfer Line Nut |
| Quantity | 1/Ea. |
| For Use With (Equipment) | SWAP-IT GC-MS Interface System for DSQ GC-MS, DSQ II GC-MS, PolarisQ GC-MS, ITQ GC-MS Only |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.