Learn More
SW-100, MedChemExpress
Product Code. 16434858
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16434858
Brand: Medchem Express HY115475
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress SW-100, a selective histone deacetylase 6 (HDAC6) inhibitor with an IC50 of 2.3 nM, shows at least 1000-fold selectivity for HDAC6 relative to all other HDAC isozymes. SW-100 displays a significantly improved ability to cross the blood-brain-barrier.
SW-100, a selective histone deacetylase 6 (HDAC6) inhibitor with an IC50 of 2.3 nM, shows at least 1000-fold selectivity for HDAC6 relative to all other HDAC isozymes. SW-100 displays a significantly improved ability to cross the blood-brain-barrier.
Specifications
Chemical Name or Material SW-100
Physical Form Solid
CAS 2126744-35-0
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C17H17ClN2O2
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : ≥ 125 mg/mL (394.60 mM)
SMILES O=C(NO)C1=CC=C(CN2CCCC3=C2C=CC(Cl)=C3)C=C1
Molecular Weight (g/mol) 316.78
Formula Weight 316.78
Percent Purity 99.33%
Grade Research