Learn More
Suxibuzone, TRC
Product Code. 30137294
Change view
Click to view available options
Quantity:
100 mg
500 mg
5 g
Unit Size:
100mg
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137294 5 g 5g
30007736 100 mg 100mg
30008301 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30137294 Supplier Toronto Research Chemicals Supplier No. TRCS8800005G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 27470-51-5
Molecular Formula C24 H26 N2 O6
Molecular Weight (g/mol) 438.47
Synonym Suxibuzone, 4-[(4-Butyl-3,5-dioxo-1,2-diphenylpyrazolidin-4-yl)methoxy]-4-oxobutanoic acid
IUPAC Name 4-[(4-butyl-3,5-dioxo-1,2-diphenylpyrazolidin-4-yl)methoxy]-4-oxobutanoic acid
SMILES CCCCC1(COC(=O)CCC(=O)O)C(=O)N(N(C1=O)c2ccccc2)c3ccccc3
Specifications
Quantity 5 g
InChI Formula InChI=1S/C24H26N2O6/c1-2-3-16-24(17-32-21(29)15-14-20(27)28)22(30)25(18-10-6-4-7-11-18)26(23(24)31)19-12-8-5-9-13-19/h4-13H,2-3,14-17H2,1H3,(H,27,28)
Formula Weight 438.1791
Percent Purity >95%
Chemical Name or Material Suxibuzone
Grade HPLC