missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SUV39H2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578581
This item is not returnable.
View return policy
Description
Positive Control: 293T, HeLa, HepG2 Predicted Reactivity: Mouse (80%), Pig (94%), Bovine (91%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The murine gene Suv39h2 encodes an H3 histone methyltransferase (HMTase) 59% identical in sequence to mouse Suv39h1. During embryogenesis, both proteins overlap in tissue expression, yet Suv39h2 transcripts are restricted to the testes in adult animals. Immunolocalization of the Suv39h2 protein during spermatogenesis indicates enrichment at the heterochromatin from the leptotene to the round spermatid stage. Moreover, Suv39h2 specifically accumulates with chromatin of the sex chromosomes, which undergo transcriptional silencing during the first meiotic prophase. Suv39h2 HMTase may also organize meiotic heterochromatin with the potential for epigenetic imprint to the male germline.
Specifications
| SUV39H2 | |
| Polyclonal | |
| Unconjugated | |
| SUV39H2 | |
| 4930507K23Rik; AA536750; D030054H19Rik; D2Ertd544e; FLJ23414; H3 K9 HMTase 2; H3-K9-HMTase 2; Histone H3-K9 methyltransferase 2; histone-lysine N-methyltransferase SUV39H2; KMT1; KMT1B; lysine N-methyltransferase 1B; RP11-2K17.2; Su; Su(var)3-9 homolog 2; suppressor of variegation 3-9 homolog 2; suppressor of variegation 3-9 homolog 2 (Drosophila); Suv39h histone methyltransferase; Suv39h2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 364785, 64707, 79723 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1.59 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q9EQQ0, Q9H5I1 | |
| SUV39H2 | |
| Recombinant protein encompassing a sequence within the center region of human SUV39H2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction