Learn More
Invitrogen™ SureCast™ Sealing Pads
Product Code. 15607968
Change view
Click to view available options
Quantity:
2 pads
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15607968 2 pads Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
Product Code. 15607968 Supplier Invitrogen™ Supplier No. HC1002
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
Leak-free, confident handcasting of your polyacrylamide gels with the Invitrogen™ SureCast™ Handcast System.
Leak-free, confident handcasting of your polyacrylamide gels with the Invitrogen™ SureCast™ Handcast System.
SureCast Sealing Pads are a replacement set of foam gaskets for use in the SureCast Handcast Station. Comes in a package of two (2) sealing pads.
  • Compatible only with the SureCast Handcast System
Recommended Storage
Room Temperature
Warranty
Up to 12 months
TRUSTED_SUSTAINABILITY
Specifications
Quantity 2 pads
For Research Use Only. Not for use in diagnostic procedures.