Learn More
Invitrogen™ SureCast™ Sealing Pads
Leak-free, confident handcasting of your polyacrylamide gels with the Invitrogen™ SureCast™ Handcast System.
Brand:  Invitrogen™ HC1002
Additional Details : Weight : 0.05000kg
Product Code. 15607968
  • 57.50 € / Pack of 2
In Stock
to see stock.
Add to basket
For Research Use Only. All usage must comply with product instructions.
Description
Description
Leak-free, confident handcasting of your polyacrylamide gels with the Invitrogen™ SureCast™ Handcast System.
SureCast Sealing Pads are a replacement set of foam gaskets for use in the SureCast Handcast Station. Comes in a package of two (2) sealing pads.
  • Compatible only with the SureCast Handcast System
Recommended Storage
Room Temperature
Services
Up to 12 months