Learn More
SMEG™ Support For 1000mL 16 Wide-necked Bottles
Product Code. 15360874
Change view
Click to view available options
:
Max. Bottle Dimensions (dia. x H): 101mm x 225mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15360874 Max. Bottle Dimensions (dia. x H): 101mm x 225mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15360874 Supplier SMEG™ Supplier No. PB1000
Please to purchase this item. Need a web account? Register with us today!
Specifications
Product Type Support
For Use With (Equipment) 1000mL 16 wide-necked bottles with GL45 screw cap
Height (Metric) 225 mm
Quantity 1 Each