Learn More
Thermo Scientific™ SUN-SRi™ 8mm Vial Crimp Seals
Product Code. 31920941 Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Silver
Septum:
PTFE/Silicone/PTFE
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Septum unitSize
31920941 Silver PTFE/Silicone/PTFE Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31920941 Supplier Thermo Scientific™ Supplier No. 201 876
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Thermo Scientific caps and vials are available in a wide variety of sizes and configurations to fit the needs of many applications.
TRUSTED_SUSTAINABILITY
Specifications
Color Silver
Material Aluminum
Pre-slit No
Septum PTFE/Silicone/PTFE
Quantity 1000 Cs.
Closure Size 8 mm
Product Type Crimp Cap