Learn More
Justrite™ Sump-to-Sump™ Drain Kit for EcoPolyBlend™ Accumulation Centers, Stainless Steel Clips, Grommets, Transfer Tube
Description
Use for applications requiring a continuous sump and for full compliance with regulations when sump capacities reach 71 gal. (270 L). Drain kit includes one pair stainless-steel joining clips, one pair rubber grommets and polytetrafluoroethylene transfer tube. It allows you to join centers together, on site, using unites 4-drum and smaller, to reach needed capacity. By joining centers together with the patented Sump-to-Sump Drain™ kit for a total 66-gallon sump capacity, you will meet EPA and UFC requirements.
Specifications
Specifications
| For Use With (Equipment) | EcoPolyBlend Accumulation Centers |