Learn More
SULT1A4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Product Code. 16828413 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16828413 20 μL 20µL
16818413 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16828413 Supplier Proteintech Supplier No. 113761AP20UL
Please to purchase this item. Need a web account? Register with us today!
Rabbit Polyclonal Antibody
Specifications
Antigen SULT1A4
Applications Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene SULT1A4
Gene Accession No. P0DMN0
Gene Alias Aryl sulfotransferase 1A3/1A4, HAST3, M PST, ST1A3/ST1A4, STM, Sulfotransferase 1A3/1A4, SULT1A3, SULT1A4, TL PST
Gene Symbols SULT1A4
Host Species Rabbit
Immunogen SULT1A4 Fusion Protein Ag1935
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 445329
Target Species Mouse, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG