missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfosuccinimidyl oleate sodium, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
Sulfosuccinimidyl oleate sodium (Sulfo-N-succinimidyl oleate sodium) is a long chain fatty acid that inhibits fatty acid transport into cells. Sulfosuccinimidyl oleate sodium is a potent and irreversible inhibitor of mitochondrial respiratory chain. Sulfosuccinimidyl oleate sodium binds the CD36 receptor on the surface of microglia. Anti-inflammatory effect.
Specifications
Specifications
| Chemical Name or Material | Sulfosuccinimidyl oleate sodium |
| CAS | 1212012-37-7 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C22H36NNaO7S |
| Quantity | 5 mg |
| Synonym | Sulfo-N-succinimidyl oleate sodium |
| Solubility Information | DMSO : 62.5 mg/mL (129.78 mM; Need ultrasonic) |
| SMILES | O=S(C(C1)C(N(OC(CCCCCCC/C=C\CCCCCCCC)=O)C1=O)=O)(O[Na])=O |
| Molecular Weight (g/mol) | 481.58 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction