missing translation for 'onlineSavingsMsg'
Learn More

Hach Lange™ SulfaVer™ 4 Sulfate Reagent Powder Pillows

Product Code. 10405972
Click to view available options
Quantity:
10 mL
100 Pack/Tests
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 10405972

Brand: Hach Lange™ 2106769

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hach Lange SulfaVer 4 Sulfate Reagent Powder Pillows, Quantity: 10mL, Physical Form: Solid

Powder Pillows make titration applications quick and easy.

  • Sealed packets ensure fresh, uncontaminated reagent every time
  • Premeasured to avoid measurement error and contamination from handling
  • Ready to use packets save time and hassle
  • Exact doses to easily monitor supply and prepare for lab procedures
  • Single-use packs for easy transportation and packing in test kits

Specifications

Format Solid
Quantity 10 mL
compliance-icons
Product Identifier
  • SulfaVer 4 Sulfate Reagent
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : citric acid, bariumchloride dihydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.