Learn More
Sulfaquinoxaline sodium salt, MedChemExpress
Product Code. 16468716
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16468716 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16468716 Supplier Medchem Express Supplier No. HYB1282A
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Sulfaquinoxaline sodium salt is an antimicrobial for veterinary use, with activity against a broad spectrum of Gram-negative and Gram-positive bacteria. Sulfaquinoxaline is used to prevent coccidiosis and bacterial infections.
Sulfaquinoxaline sodium salt is an antimicrobial for veterinary use, with activity against a broad spectrum of Gram-negative and Gram-positive bacteria. Sulfaquinoxaline is used to prevent coccidiosis and bacterial infections.
Specifications
Chemical Name or Material Sulfaquinoxaline sodium salt
Physical Form Solid
CAS 967-80-6
Color Light Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C14H11N4NaO2S
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 100 mg/mL (310.25 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=S(C1=CC=C(N)C=C1)([N-]C2=NC3=CC=CC=C3N=C2)=O.[Na+]
Molecular Weight (g/mol) 322.32
Formula Weight 322.32
Percent Purity 98.0%
Grade Research