Learn More
Sulfapyridine, MedChemExpress
Product Code. 16482089
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 16482089
Brand: Medchem Express HYB0212
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Sulfapyridine, a major metabolite of Sulfasalazine, is a sulfonamide antibiotic agent. Sulfapyridine inhibits recombinant P. carinii dihydropteroate synthetase (DHPS) with an IC50 of 0.18 μM. Sulfapyridine has antibacterial, anti-inflammatory and anti-rheumatic activities.
Sulfapyridine, a major metabolite of Sulfasalazine, is a sulfonamide antibiotic agent. Sulfapyridine inhibits recombinant P. carinii dihydropteroate synthetase (DHPS) with an IC50 of 0.18 μM. Sulfapyridine has antibacterial, anti-inflammatory and anti-rheumatic activities.
Specifications
Chemical Name or Material Sulfapyridine
Physical Form Solid
CAS 144-83-2
Color Off-White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C11H11N3O2S
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 60 mg/mL (240.68 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=S(C1=CC=C(N)C=C1)(NC2=NC=CC=C2)=O
Molecular Weight (g/mol) 249.29
Formula Weight 249.29
Percent Purity 98.86%
Grade Research