Learn More
Sulfamethoxazole, MedChemExpress
Product Code. 16492096
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492096 500 mg 500mg
16418236 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16492096 Supplier Medchem Express Supplier No. HYB0322
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket
MedChemExpress Sulfamethoxazole (Ro 4-2130) is a sulfonamide bacteriostatic antibiotic, used for bacterial infections. Sulfonamides is a competitive antagonists of para-aminobenzoic acid (PABA).
Sulfamethoxazole (Ro 4-2130) is a sulfonamide bacteriostatic antibiotic, used for bacterial infections. Sulfonamides is a competitive antagonists of para-aminobenzoic acid (PABA).
Specifications
Chemical Name or Material Sulfamethoxazole
Physical Form Solid
CAS 723-46-6
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C10H11N3O3S
For Use With (Application) COVID-19-immunoregulation
Synonym Ro 4-2130
Solubility Information DMSO : ≥ 100 mg/mL (394.82 mM) ∣H2O : 0.1 mg/mL (0.39 mM; Need ultrasonic)
SMILES O=S(C1=CC=C(N)C=C1)(NC2=NOC(C)=C2)=O
Molecular Weight (g/mol) 253.28
Formula Weight 253.28
Percent Purity 99.93%
Grade Research