Learn More
Sulfadimethoxine D6 (2,6-dimethoxy D6) 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Sulfadimethoxine D6 (2,6-dimethoxy D6) |
| CAS | 73068-02-7 |
| Solvent or Matrix | Acetonitrile |
| Molecular Formula | C12 2H6 H8 N4 O4 S |
| Quantity | 1 mL |
| Synonym | Sulfadimethoxine D6 (2,6-dimethoxy D6) |
| InChI Formula | InChI=1S/C12H14N4O4S/c1-19-11-7-10(14-12(15-11)20-2)16-21(17,18)9-5-3-8(13)4-6-9/h3-7H,13H2,1-2H3,(H,14,15,16)/i1D3,2D3 |
| SMILES | [2H]C([2H])([2H])Oc1cc(NS(=O)(=O)c2ccc(N)cc2)nc(OC([2H])([2H])[2H])n1 |
| IUPAC Name | 4-amino-N-[2,6-bis(trideuteriomethoxy)pyrimidin-4-yl]benzenesulfonamide |
| Molecular Weight (g/mol) | 316.37 |
| Show More |