Learn More
Sudan III-d6, TRC
Product Code. 30423790
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30423790 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30423790 Supplier Toronto Research Chemicals Supplier No. TRCS688902100MG
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Sudan III-D6
CAS 1014689-17-8
Molecular Formula C22 2H6 H10 N4 O
Quantity 100 mg
Synonym Sudan 3 D6 (naphthyl D6), 2-Naphthalen-3,4,5,6,7,8-d6-ol','1-[2-[4-(2-phenyldiazenyl)phenyl]diazenyl]-, 1-[2-[4-(2-Phenyldiazenyl)phenyl]diazenyl]-2-naphthalen-3,4,5,6,7,8-d6-ol (ACI), 2-Naphthalen-3,4,5,6,7,8-d6-ol','1-[2-[4-(2-phenyldiazenyl)phenyl]diazenyl]-
InChI Formula InChI=1S/C22H16N4O/c27-21-15-10-16-6-4-5-9-20(16)22(21)26-25-19-13-11-18(12-14-19)24-23-17-7-2-1-3-8-17/h1-15,27H/i4D,5D,6D,9D,10D,15D
SMILES [2H]c1c([2H])c([2H])c2c(N=Nc3ccc(cc3)N=Nc4ccccc4)c(O)c([2H])c([2H])c2c1[2H]
IUPAC Name 3,4,5,6,7,8-hexadeuterio-1-[(E)-[4-[(E)-phenyldiazenyl]phenyl]diazenyl]naphthalen-2-ol
Molecular Weight (g/mol) 358.43
Formula Weight 358.1701