Learn More
Sudan III-d6, TRC
Specifications
Specifications
| Chemical Name or Material | Sudan III-D6 |
| CAS | 1014689-17-8 |
| Molecular Formula | C22 2H6 H10 N4 O |
| Quantity | 10 mg |
| Synonym | Sudan 3 D6 (naphthyl D6), 2-Naphthalen-3,4,5,6,7,8-d6-ol','1-[2-[4-(2-phenyldiazenyl)phenyl]diazenyl]-, 1-[2-[4-(2-Phenyldiazenyl)phenyl]diazenyl]-2-naphthalen-3,4,5,6,7,8-d6-ol (ACI), 2-Naphthalen-3,4,5,6,7,8-d6-ol','1-[2-[4-(2-phenyldiazenyl)phenyl]diazenyl]- |
| InChI Formula | InChI=1S/C22H16N4O/c27-21-15-10-16-6-4-5-9-20(16)22(21)26-25-19-13-11-18(12-14-19)24-23-17-7-2-1-3-8-17/h1-15,27H/i4D,5D,6D,9D,10D,15D |
| SMILES | [2H]c1c([2H])c([2H])c2c(N=Nc3ccc(cc3)N=Nc4ccccc4)c(O)c([2H])c([2H])c2c1[2H] |
| IUPAC Name | 3,4,5,6,7,8-hexadeuterio-1-[(E)-[4-[(E)-phenyldiazenyl]phenyl]diazenyl]naphthalen-2-ol |
| Molecular Weight (g/mol) | 358.43 |
| Formula Weight | 358.1701 |