missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SUCLG2 Polyclonal Antibody

Description
Peptide sequence: AKINFDDNAE FRQKDIFAMD DKSENEPIEN EAAKYDLKYI GLDGNIACFV Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
This gene encodes a GTP-specific beta subunit of succinyl-CoA synthetase. Succinyl-CoA synthetase catalyzes the reversible reaction involving the formation of succinyl-CoA and succinate. Alternate splicing results in multiple transcript variants. Pseudogenes of this gene are found on chromosomes 5 and 12.
Specifications
Specifications
| Antigen | SUCLG2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | SUCLG2 |
| Gene Accession No. | Q96I99 |
| Gene Alias | AF171077; AW556404; D6Wsu120e; GBETA; G-BETA; G-SCS; GTPSCS; GTP-specific succinyl-CoA synthetase beta subunit; GTP-specific succinyl-CoA synthetase subunit beta; hypothetical protein LOC537713; SCS-betaG; Succinate--CoA ligase [GDP-forming] subunit beta, mitochondrial; succinate-CoA ligase GDP-forming beta subunit; succinate-CoA ligase GDP-forming subunit beta; succinate-CoA ligase, GDP-forming, beta subunit; succinate-Coenzyme A ligase, GDP-forming, beta subunit; succinyl-CoA ligase [GDP-forming] subunit beta, mitochondrial; succinyl-CoA ligase, GDP-forming, beta chain, mitochondrial; succinyl-CoA synthetase beta-G chain; succinyl-CoA synthetase, beta-G chain; Suclg2 |
| Gene Symbols | SUCLG2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?