Learn More
STEMCELL Technologies SU5402
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Description
- SU5402 is a potent and selective inhibitor of vascular endothelial growth factor receptor 2 (VEGFR-2), fibroblast growth factor receptor 1 (FGFR-1), and platelet-derived growth factor receptor beta (PDGFRB) with IC50 = 0.02, 0.03, and 0.51 μM, respectively (Sun et al.)
Specifications
Specifications
| CAS | 215543-92-3 |
| Quantity | 5 mg |
| Molecular Formula | C17H16N2O3 |
| Purity | ≥95% |