missing translation for 'onlineSavingsMsg'
Learn More

SU1498, MedChemExpress

Product Code. 16461799
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16461799

Brand: Medchem Express HY19326

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SU1498 (AG 1498) is a selective inhibitor of the VEGFR2; inhibits Flk-1 with an IC50 of value of 700 nM.

SU1498 (AG 1498) is a selective inhibitor of the VEGFR2

Specifications

Chemical Name or Material SU1498
CAS 168835-82-3
Color White
Purity Grade Notes Research
Molecular Formula C25H30N2O2
Quantity 1 mg
Synonym AG 1498 Tyrphostin SU 1498
Solubility Information DMSO : 100 mg/mL (256.07 mM; Need ultrasonic)
SMILES O=C(NCCCC1=CC=CC=C1)/C(C#N)=C/C2=CC(C(C)C)=C(O)C(C(C)C)=C2
Molecular Weight (g/mol) 390.52
Formula Weight 390.52
Percent Purity 98.37%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.