Learn More
SU14813 maleate, MedChemExpress
Product Code. 16494008
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16494008
Brand: Medchem Express HY10501A
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress SU14813 maleate is a multi-targeted receptor tyrosine kinases inhibitor with IC50s of 50, 2, 4, 15 nM for VEGFR2, VEGFR1, PDGFRβ and KIT.
SU14813 maleate is a multi-targeted receptor tyrosine kinases inhibitor with IC50s of 50, 2, 4, 15 nM for VEGFR2, VEGFR1, PDGFRβ and KIT.
Specifications
Chemical Name or Material SU14813 maleate
CAS 849643-15-8
Color Orange
Purity Grade Notes Research
Molecular Formula C27H31FN4O8
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (179.03 mM)
SMILES O=C(NC[C@@H](CN1CCOCC1)O)C2=C(NC(/C=C3C(NC4=C\3C=C(C=C4)F)=O)=C2C)C.O=C(/C=C\C(O)=O)O
Molecular Weight (g/mol) 558.56
Formula Weight 558.56
Percent Purity 99.95%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease