Learn More
Cole-Parmer™ Stuart™ Wall Supports
Product Code. 12964447
Click to view available options
:
For use with Bibbyjet Pro Pipette Controller PC2000/2001
Product Code. 12964447
Brand: Cole-Parmer™ 9995555
Please to purchase this item. Need a web account? Register with us today!
For use with Bibbyjet Pro™ Pipette Controller PC2000/2001
  • Lightweight and ergonomic
  • Powerful enough to fill a 50 mL pipette in just 10s
  • Battery sufficient to give a full day's work before recharging - LED display shows when battery is low
  • Pipette holder fully autoclavable
  • Venting feature allows air sucked from the pipette to be discharged from the front of the unit, avoiding corrosion of internal mechanisms
Specifications
Type Wall Support
For Use With (Equipment) Bibbyjet Pro Pipette Controller PC2000/2001