Learn More
Cole-Parmer™ Stuart™ Temperature Controller
Product Code. 12952600
Click to view available options
Temperature Range (Metric):
0°C to 630°C
Unit Size:
Each
Product Code. 12952600
Brand: Cole-Parmer™ 1620981
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Length (Metric) 240 mm
Temperature Range (Metric) 0°C to 630°C
Width (Metric) 165 mm
Dimensions (L x W x H) 240 x 165 x 140mm
Frequency 50/60 Hz
Type Temperature Controller
Voltage 120/230 V