Learn More
Cole-Parmer™ Stuart™ Snap Tapes
Click to view available options
:
For use with Bibbyjet Pro Pipette Controller PC2000/2000
Beschreibung
- Lightweight and ergonomic
- Powerful enough to fill a 50 mL pipette in just 10 s
- Battery sufficient to give a full day's work before recharging - LED display shows when battery is low
- Pipette holder fully autoclavable
- Venting feature allows air sucked from the pipette to be discharged from the front of the unit, avoiding corrosion of internal mechanisms
Spezifikation
Spezifikation
| Autoclavable | Autoclavable |
| Type | Snap Tapes |
| For Use With (Equipment) | Bibbyjet Pro Pipette Controller PC2000/2001 |
| Dimensions (L x W x H) | 12 x 65mm |