Learn More
Cole-Parmer™ Stuart™ Rotisserie for Hybridization Oven
Product Code. 11406568
Click to view available options
No. of Positions:
2
7
Unit Size:
Each
Product Code. 11406568
Brand: Cole-Parmer™ 3945495
Only null left
Add to basket
Add to basket
Stuart™ Rotisserie for Hybridization Oven for Cole-Parmer™ INC-200D-H Series Stuart Hybridization Incubator/Shakers.
  • Choose from a wide range of accessories for your Cole-Parmer hybridization incubator.
  • Select from differnt capacity rotisseries and bottles to suit your specific applicaiton.
Specifications
Product Type Rotisserie
For Use With (Equipment) Hybridization bottles: 39454-97/-99
No. of Positions 7