Learn More
Cole-Parmer™ Stuart™ Pipette Controller Adapters
Click to view available options
For Use With (Equipment):
Bibbyjet Pro Pipette Controller PC2000/2001
Unit Size:
Each
Description
- Silicone Adapter
- For use with PC2000 Bibbyjet Pro Pipette Controller
- Lightweight and ergonomic
- Powerful enough to fill a 50mL pipette in just 10s
- Battery sufficient to give a full day's work before recharging - LED display shows when battery is low
- Pipette holder fully autoclavable
- Venting feature allows air sucked from the pipette to be discharged from the front of the unit, avoiding corrosion of internal mechanisms
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | Silicone |
| Type | Pipette Controller Adapter |
| For Use With (Equipment) | Bibbyjet Pro Pipette Controller PC2000/2001 |