Learn More
Cole-Parmer™ Stuart™ Clear Protective Discs
Click to view available options
Quantity:
5 Pk.
Unit Size:
Pack of 5
Description
- Protection Disc, Translucent/Opaque
Specifications
Specifications
| Product Type | Protective Discs |
| Color | Clear |
| Quantity | 5 Pk. |
| For Use With (Equipment) | Colony Counter |