Learn More
Invitrogen™ eBioscience™ Streptavidin FITC Conjugate
Commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.
Brand: Invitrogen™ 11-4317-87
Additional Details : Weight : 0.09500kg
Alert:
Contains Azide
Specifications
0.5 mg/mL | |
Liquid | |
Streptavidin Conjugate (Fluorescent) | |
FITC (Fluorescein) |
FITC (Fluorescein) | |
eBioscience™ | |
Biotin |
For Research Use Only