missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brand™ Straight Tubing Connector
Shop All Brand Products
Click to view available options
Overall Length (Metric):
66 mm
Unit Size:
Pack of 10
Description
- Good chemical resistance and mechanical properties
- Provided in packs of 10
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | Polypropylene |
| Overall Length (Metric) | 66 mm |
| For Use With | Tubing with inner diameter of 13-15mm |
| Packaging Quantity | 10/Pk. |
| Color | Gray |
| Product Type | Tubing Connector |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction