Learn More
BRAND™ Straight Burette Stopcock for Burette
Product Code. 10400683 Shop All Brand Products
Click to view available options
For Use With (Equipment):
2 to 10 mL Burette
25 mL Burette
50 mL Burette
Unit Size:
Each
This item is not returnable. View return policy
Product Code. 10400683
Brand: BRAND™ 81817
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Straight Burette Stopcock for Burette includes an interchangeable glass key.
  • Stopcock housing with precision Borosilicate Glass 3.3 burette tip
  • Interchangeable borosilicate glass key with retention device
  • Stopcock size 3NS/12
TRUSTED_SUSTAINABILITY
Specifications
Type Burette Stopcock
Material Borosilicate Glass (3.3)
For Use With (Equipment) 25 mL Burette
Includes Glass Key