Learn More
BRAND™ Straight Burette Stopcock for Burette
Product Code. 10365471 Shop All Brand Products
Click to view available options
For Use With (Equipment):
2 to 10 mL Burette
25 mL Burette
50 mL Burette
Unit Size:
Each
Product Code. 10365471
Brand: BRAND™ 81815
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Straight Burette Stopcock for Burette includes an interchangeable glass key.
  • Stopcock housing with precision Borosilicate Glass 3.3 burette tip
  • Interchangeable borosilicate glass key with retention device
  • Stopcock size 3NS/12
TRUSTED_SUSTAINABILITY
Specifications
Type Burette Stopcock
Material Borosilicate Glass (3.3)
For Use With (Equipment) 2 to 10 mL Burette
Includes Glass Key