missing translation for 'onlineSavingsMsg'
Learn More

STRADA, Rabbit anti-Human, Polyclonal Antibody, Abnova™

Product Code. 16175389
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16175389 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16175389 Supplier Abnova Supplier No. H00092335D01P.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Sequence: MSFLTNDASSESIASFSKQEVMSSFLPEGGCYELLTVIGKGFEDLMTVNLARYKPTGEYVTVRRINLEACSNEMVTFLQGELHVSKLFNHPNIVPYRATFIADNELWVVTSFMAYGSAKDLICTHFMDGMNELAIAYILQGVLKALDYIHHMGYVHRSVKASHILISVDGKVYLSGLRSNLSMISHGQRQRVVHDFPKYSVKVLPWLSPEVLQQNLQGYDAKSDIYSVGITACELANGHVPFKDMPATQMLLEKLNGTVPCLLDTSTIPAEELTMSPSRSVANSGLSDSLTTSTPRPSNGDSPSHPYHRTFSPHFHHFVEQCLQRNPDARYPCWPGPGLRESRGCSGG

Specifications

Antigen STRADA
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene STE20-related kinase adaptor alpha
Gene Accession No. NM_153335
Gene Alias FLJ90524/LYK5/NY-BR-96/PMSE/STRAD/Stlk
Gene Symbols STRADA
Host Species Rabbit
Immunogen STRADA (NP_699166.2, 1 a.a. ∼ 348 a.a) full-length human protein.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92335
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.