missing translation for 'onlineSavingsMsg'
Learn More

STO-609, MedChemExpress

Product Code. 16427726
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16427726

Brand: Medchem Express HY19805

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress STO-609 is a selective and cell-permeable inhibitor of the Ca2+/calmodulin-dependent protein kinase kinase (CaM-KK), with Ki values of 80 and 15 ng/mL for recombinant CaM-KKα and CaM-KKβ, respectively. STO-609 inhibits AMP-activated protein kinase kinase (AMPKK) activity in HeLa cell lysates with an IC50 ∼0.02 g/ml.

STO-609 is a selective and cell-permeable inhibitor of the Ca2+/calmodulin-dependent protein kinase kinase (CaM-KK), with Ki values of 80 and 15 ng/mL for recombinant CaM-KKα and CaM-KKβ, respectively. STO-609 inhibits AMP-activated protein kinase kinase (AMPKK) activity in HeLa cell lysates with an IC50 ∼0.02 g/ml.

Specifications

Chemical Name or Material STO-609
CAS 52029-86-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C19H10N2O3
Quantity 100 mg
Solubility Information DMSO : 6.25 mg/mL (19.89 mM; ultrasonic and warming and heat to 80°C) ∣DMF : 5.56 mg/mL (17.69 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C1=C2C3=C(C4=NC5=CC=CC=C5N4C(C3=CC=C2)=O)C=C1)O
Molecular Weight (g/mol) 314.29
Formula Weight 314.29
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.