missing translation for 'onlineSavingsMsg'
Learn More

Stiripentol, MedChemExpress

Product Code. p-200100848
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16464876

Brand: Medchem Express HY103392

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Stiripentol (STP) is an anticonvulsant agent, which can inhibit N-demethylation of CLB to NCLB mediatated by CYP3A4 (noncompetitively) and CYP2C19 (competitively) with Ki of 1.59±0.07 and 0.516±0.065 μM and IC50 of 1.58 and 3.29 μM, respectively.

Stiripentol (STP) is an anticonvulsant agent, which can inhibit N-demethylation of CLB to NCLB mediatated by CYP3A4 (noncompetitively) and CYP2C19 (competitively) with Ki of 1.59±0.07 and 0.516±0.065 μM and IC50 of 1.58 and 3.29 μM, respectively.

Specifications

Chemical Name or Material Stiripentol
CAS 49763-96-4
Color White
Purity Grade Notes Research
Molecular Formula C14H18O3
Quantity 100 mg
Synonym BCX2600
Solubility Information DMSO : 150 mg/mL (640.23 mM; Need ultrasonic and warming)
SMILES CC(C)(C)C(O)/C=C/C1=CC=C(OCO2)C2=C1
Molecular Weight (g/mol) 234.29
Formula Weight 234.29
Percent Purity 99.97%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.