Learn More
Stiripentol, MedChemExpress
Product Code. 16464876
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16464876
Brand: Medchem Express HY103392
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Stiripentol (STP) is an anticonvulsant agent, which can inhibit N-demethylation of CLB to NCLB mediatated by CYP3A4 (noncompetitively) and CYP2C19 (competitively) with Ki of 1.59±0.07 and 0.516±0.065 μM and IC50 of 1.58 and 3.29 μM, respectively.
Stiripentol (STP) is an anticonvulsant agent, which can inhibit N-demethylation of CLB to NCLB mediatated by CYP3A4 (noncompetitively) and CYP2C19 (competitively) with Ki of 1.59±0.07 and 0.516±0.065 μM and IC50 of 1.58 and 3.29 μM, respectively.
Specifications
Chemical Name or Material Stiripentol
CAS 49763-96-4
Color White
Purity Grade Notes Research
Molecular Formula C14H18O3
Quantity 100 mg
Synonym BCX2600
Solubility Information DMSO : 150 mg/mL (640.23 mM; Need ultrasonic and warming)
SMILES CC(C)(C)C(O)/C=C/C1=CC=C(OCO2)C2=C1
Molecular Weight (g/mol) 234.29
Formula Weight 234.29
Percent Purity 99.97%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation