missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STIP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583700
This item is not returnable.
View return policy
Description
Immunogen sequence: YARIGNSYFK EEKYKDAIHF YNKSLAEHRT PDVLKKCQQA EKILKEQERL AYINPDLALE EKNKGNECFQ KGDYPQAMKH YTEAIKRNPK DAKLYS Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 98%.
Mediates the association of the molecular chaperones HSC70 and HSP90 (HSPCA and HSPCB).
Specifications
| STIP1 | |
| Polyclonal | |
| Unconjugated | |
| STIP1 | |
| epididymis secretory sperm binding protein Li 94n; HEL-S-94n; HOP; hsc70/Hsp90-organizing protein; Hsp70/Hsp90 organizing protein; Hsp70/Hsp90-organizing protein; IEF SSP 3521; IEF-SSP-3521; mSTI1; NY-REN-11 antigen; P60; Renal carcinoma antigen NY-REN-11; STI1; STI1L; Stip1; stress induced phosphoprotein 1; stress-induced phosphoprotein 1; stress-induced-phosphoprotein 1; stress-induced-phosphoprotein 1 (Hsp70/Hsp90-organizing protein); stress-inducible protein; transformation-sensitive protein IEF SSP 3521 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10963 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P31948 | |
| STIP1 | |
| Recombinant protein corresponding to Human STIP1. Recombinant protein control fragment (Product #RP-97460). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction