missing translation for 'onlineSavingsMsg'
Learn More

STING agonist-1, MedChemExpress

Product Code. 16427676
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16427676

Brand: Medchem Express HY19711

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress STING agonist-1 (G10) is human-specific STING agonist that elicits antiviral activity against emerging Alphaviruses. G10 potently blocks replication of Alphavirus species Venezuelan Equine Encephalitis Virus (VEEV) with IC90 of 24.57 μM.

STING agonist-1 (G10) is human-specific STING agonist that elicits antiviral activity against emerging Alphaviruses. G10 potently blocks replication of Alphavirus species Venezuelan Equine Encephalitis Virus (VEEV) with IC90 of 24.57 μM.

Specifications

Chemical Name or Material STING agonist-1
Physical Form Solid
CAS 702662-50-8
Color Light Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C21H16ClFN2O3S
For Use With (Application) COVID-19-anti-virus
Synonym G10
Solubility Information DMSO : 100 mg/mL (232.08 mM; Need ultrasonic)
SMILES O=C(C1=CC=C(SCC(N2CC3=C(F)C=CC=C3Cl)=O)C2=C1)NCC4=CC=CO4
Molecular Weight (g/mol) 430.88
Formula Weight 430.88
Percent Purity 99.54%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.