Learn More
ibidi sticky-Slide Tissue
Description
- A self-adhesive chamber for customized cell-based assays and microscopy
- Self-adhesive chamber for tissue sections and microscopy
- Sterilized
- Individually packed
Specifications
Specifications
| Type | Chamber Slide |
| For Use With (Application) | Cell-based Assays, Microscopy |
| No. per Case | 90/Cs. |
| No. per Pack | 1 Ea. |
| Sterility | Sterile |