missing translation for 'onlineSavingsMsg'
Learn More

STF-62247, MedChemExpress

Product Code. p-200100626
Encompass_Preferred
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
This item is not returnable. View return policy

Product Code. 16449706

Brand: Medchem Express HY100746

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress STF-62247 is an autophagy inducer that selectively cytotoxic to VHL-deficient renal cell carcinoma (IC50 of 0.625 μM and 16 μM in RCC4 and RCC4/VHL cells, respectively).

STF-62247 is an autophagy inducer that selectively cytotoxic to VHL-deficient renal cell carcinoma (IC50 of 0.625 μM and 16 μM in RCC4 and RCC4/VHL cells, respectively).

Specifications

Chemical Name or Material STF-62247
Physical Form Solid
CAS 315702-99-9
Color Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C15H13N3S
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 50 mg/mL (187.02 mM; Need ultrasonic)
SMILES CC1=CC(NC2=NC(C3=CC=NC=C3)=CS2)=CC=C1
Molecular Weight (g/mol) 267.35
Formula Weight 267.35
Percent Purity 98.99%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.