Learn More
Brady™ Sterilization Indicating Labels
Product Code. 17110191 Shop All Brady Products
Change view
Click to view available options
Height (English):
1.5 in.
2 in.
Unit Size:
Pack of 2000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (English) Unit Size
17110191 2 in. Pack of 2000
17100191 1.5 in. Pack of 2000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17110191 Supplier Brady™ Supplier No. 149273
  • ONLINE EXCLUSIVE PRICE
    235.00 €
    205.47 € / Pack of 2000
    Save 29.53 €
    12% Off
View all promotions
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Brady™ Sterilization Indicating Labels are steam sterilization indicating polypropylene labels with preprinted headers that displays STERILIZED when exposed to autoclave conditions.
  • Ideal for use in sample tracking, inventory management, general tracking, and identification of lab materials that require sterilization
  • Economical solution for identifying vials, tubes, racks and flasks
  • Adheres to stainless steel, glass and polypropylene
  • Withstands temperatures from -320.8 to 249.8° F (-196 to 121°C )
Compliance
RoHS directive 2011/65/EU
Specifications
Autoclavable Autoclavable
Adhesive Type Acrylic, Permanent
Coating None
Color White
Finish Matte
Format Die-cut
Material Polypropylene
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Component and equipment labelling, General identification, Laboratory labelling, Temperature indicating labelling
For Use With (Equipment) BBP81, i5100, i6100, i7100, i7500, PR Plus, IP Series, THT 76 mm Core Printers
Labels per Row 1
Min. Operating Temperature (Metric) -80°C
Model THT-137-7425-2-AC
No. per Pack 2000 Label(s) / Roll
Quantity 2000/Pk.
Shape Rectangle
Size 50.80 mm (W) x 25.40 mm (H)
Temperature Range (English) -112°F to +158°F
Temperature Range (Metric) -80°C to +70°C
Thickness (English) 0.004 in.
Thickness (Metric) 0.102 mm
Dimensions (L x W) 50.80 mm (W) x 25.40 mm (H)
Length (English) 2 in.
Width (English) 1 in.
Width (Metric) 50.80 mm
Width (English) Printable Area 50.80 mm
Width (Metric) Printable Area 50.80 mm
Height (English) 2 in.
Height (English) Printable Area 0.75 in.
Height (Metric) 25.40 mm
Height (Metric) Printable Area 19.05 mm
Alternate Part Number THT-137-7425-2-AC