Learn More
SP Bel-Art™ Sterileware Volumetric Sampling Spoons
Suitable for use with powders, granules, liquids, gels or suspensions, these sterile spoons allow set volumes to be accurately measured and sampled
Brand:  SP Bel-Art™ F36943-0005
Additional Details : Weight : 85.00000kg
 View more versions of this product
Product Code. 16826916
  • 147.00 € / Pack of 100
Estimated Shipment: 04-07-2024
to see stock.
Add to basket
Description
Description
  • Leveling to top of bowl provides accurately measured samples
  • Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
  • High density polyethylene (HDPE) spoons heat sealed in individual polyethylene bags
  • Gamma irradiated sterile (SAL 10^-6)
  • Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization
Specifications
Specifications
5 mL
100/Pk.
16.5 cm