missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B Braun™ SterilContainer™ 2000 Mini-Size Container Lids
Click to view available options
Color:
Blue
Red
Unit Size:
Each
Description
- Low operating cost and typical 10+ year service life
- Simple, fast and efficient with lightweight aluminum and ergonomic handles
- Clear identification using colored lids, indicator labels, identification labels and barcoding
- Lid design of one-piece construction for easier maintenance and lower repair costs
Specifications
Specifications
| Color | Red |
| For Use With (Application) | Sterilization, transport, storage |
| For Use With (Equipment) | Mini-Size SterilContainers™ |
| Product Type | Container Lid |
| Material | Aluminum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction