missing translation for 'onlineSavingsMsg'
Learn More

B Braun™ SterilContainer™ 2000 Mini-Size Container Lids

Product Code. 10469934
Click to view available options
Color:
Blue
Red
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10469934

Brand: B Braun™ JK170

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Helps ensure the proper storage, transportation and sterilization of surgical instruments.

  • Low operating cost and typical 10+ year service life
  • Simple, fast and efficient with lightweight aluminum and ergonomic handles
  • Clear identification using colored lids, indicator labels, identification labels and barcoding
  • Lid design of one-piece construction for easier maintenance and lower repair costs

Specifications

Color Red
For Use With (Application) Sterilization, transport, storage
For Use With (Equipment) Mini-Size SterilContainers™
Product Type Container Lid
Material Aluminum
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.