Learn More
STEMCELL Technologies StemSpan™-XF
Product Code. 17127461
Encompass
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17127461 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17127461 Supplier STEMCELL Technologies Supplier No. 1000073
Please to purchase this item. Need a web account? Register with us today!
Xeno-free and serum-free medium for culture and expansion of human hematopoietic cells
  • StemSpan™-XF has been developed for the in vitro culture and expansion of human hematopoietic cells, when the appropriate growth factors and supplements are added
  • This allows users the flexibility to prepare medium that meets their requirements
  • StemSpan™-XF contains pre-tested human-derived and recombinant human (rh) proteins
  • Using appropriate StemSpan™ Expansion Supplements, StemSpan™-XF may be used to expand CD34+ cells isolated from human cord blood, mobilized peripheral blood, or bone marrow samples, or to expand and differentiate lineage-committed progenitor cells to generate populations of erythroid, myeloid, or megakaryocyte progenitor cells
Specifications
Cell Type Hematopoietic Stem and Progenitor Cells
For Use With (Application) Cell Culture, Expansion
Quantity 500 mL