missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies StemSpan™ Myeloid Expansion Supplement II (100X)

Product Code. 17198051
Encompass
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17198051 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17198051 Supplier STEMCELL Technologies Supplier No. 02694

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Serum-free culture supplement for expansion and differentiation of human monocytes

  • StemSpan™ Myeloid Expansion Supplement II (100X) contains a combination of recombinant human cytokines formulated to selectively promote the expansion and differentiation of human myeloid progenitor cells, generating large numbers of CD14+ monocytes from CD34+ cells isolated from human cord blood (CB) samples
  • StemSpan™ Myeloid Expansion Supplement II (100X) is intended for use in combination with any of the following StemSpan™ media: StemSpan™ SFEM (Catalog #09600)
  • StemSpan™ SFEM II (Catalog #09605)
  • StemSpan™-XF (Catalog #100-0073)
  • StemSpan™-ACF Without Phenol Red (Catalog #100-0130)When added to serum-free medium, StemSpan™ Myeloid Expansion Supplement II typically promotes the production of hundreds of CD14+ monocytes per input CD34+ cell in 14-day liquid cultures initiated with CD34+ human CB cells
  • See data tab for more details

Specifications

Cell Type Hematopoietic Stem and Progenitor Cells, Monocytes, Myeloid Cells
For Use With (Application) Cell Culture, Differentiation, Expansion
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.